Learn More
ALB, Mouse, Clone: AL-01, Abnova™
Mouse monoclonal antibody raised against native ALB.
Marca: Abnova MAB3598.100ug
Descripción
Albumin is a soluble, monomeric protein which comprises about one-half of the blood serum protein. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular unglycosylated serum protein of molecular weight 65,000. Albumin is synthesized in the liver as preproalbumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough endoplasmic reticulum. The product, proalbumin, is in turn cleaved in the Golgi vesicles to produce the secreted albumin. [provided by RefSeq
Especificaciones
ALB | |
Monoclonal | |
1 mg/mL | |
Mouse monoclonal antibody raised against native ALB. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
DKFZp779N1935/PRO0883/PRO0903/PRO1341 | |
Mouse | |
100 μg | |
Primary | |
This antibody reacts only with human serum albumin, a 65-67 KDa monomeric proteinin human blood plasma; it is produced in liver.No cross-reactivity was observed with other serum proteins. | |
Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Immunohistochemistry, Radioimmune Assays (RIA), Western Blot | |
AL-01 | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user. | |
ALB | |
ALB | |
Native purified human ALB. | |
RUO | |
213 | |
Human | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.