missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Leptin/OB Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Marca: R&D Systems MAB4981-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Leptin/OB Monoclonal antibody specifically detects Leptin/OB in Mouse samples. It is validated for ELISA, NeutralizationEspecificaciones
Leptin/OB | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
FLJ94114, LEP, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), OB, Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS | |
E. coli-derived mouse Leptin/OB protein, Val22-Cys167, Accession # Q544U0 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG |
ELISA, Neutralization | |
2299A | |
ELISA, Neutralization 0.1-1 ug/mL | |
Q544U0 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
3952 | |
Mouse | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido