Learn More
WTW™ MultiLine™ Multi 3620 IDS Multi-parameter Portable Meter
Solve multiple measuring tasks at once and save time and costs. WTW™ MultiLine™ Multi 3620 IDS Multi-parameter Portable Meters feature two universal measuring channels for measuring pH, ORP, dissolved oxygen and/or turbidity simultaneously.
1605.00€ - 3360.00€
Especificaciones
Canales 2
Tipo de pantalla Pantalla LCD gráfica en color, retroiluminada
Clasificación IP IP 67
Memoria Manual 500/10000 automático
Fuente de alimentación Fuente de alimentación de intervalo amplio, 4 pilas NiMH de 1,2 V o a través de mini USB-B
Ver más especificaciones
Productos 2
Código de producto Marca Modelo Precio Cantidad y disponibilidad  
Código de producto Marca Modelo Precio Cantidad y disponibilidad  
15568595
Ver documentos Detalles de la promoción
WTW™
2FD560
Medidor portátil multiparamétrico MultiLine™ Multi 3620 IDS
1605.00€
cada uno
15520476
Ver documentos
WTW™
2FD56G
Conjunto G de medidor portátil multiparamétrico MultiLine™ Multi 3620 IDS
3360.00€
cada uno
Descripción
Descripción
  • Wireless ready
  • Simultaneous display of all measured values
  • Digital sensor recognition
  • Two galvanically isolated measuring inputs
  • Dual – 2 channels, 4 parameters
  • Galvanic isolation of the sensor inputs prevents interferences of measuring signals
  • Use the new wireless modules together with new IDS plug head sensors, be independent from cables and measure even at hardly accessible locations
  • Efficient battery or mains supply, you will be able to measure cost-effectively in the field and in the lab
  • Data transfer either takes place via USB stick or via an USB connection directly to a computer
  • Color graphic display offers measured values as well as additional information such as sensor position, CMC and QSC
  • Traceability of the individual measured values can be supported by activating the user administration
Scope of delivery includes the MultiLab™ Importer to import data via Excel™ as well as the MultiLab™ User (user administration).
Especificaciones
Especificaciones
2
IP 67
Fuente de alimentación de intervalo amplio, 4 pilas NiMH de 1,2 V o a través de mini USB-B
Pantalla LCD gráfica en color, retroiluminada
Manual 500/10000 automático