missing translation for 'onlineSavingsMsg'
Learn More

N-(2,6-Dichlorophenyl)-2-fluorobenzamide, 97%, Thermo Scientific™

Product Code. 15455948
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15455948 250 mg 250mg
15445948 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15455948

Brand: Thermo Scientific Alfa Aesar H59580.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304884-73-9
Molecular Formula C13H8Cl2FNO
Molecular Weight (g/mol) 284.111
MDL Number MFCD00578380
InChI Key CSQKIRJRBCAPIY-UHFFFAOYSA-N
Synonym n-2,6-dichlorophenyl-2-fluorobenzamide, n-2,6-dichlorophenyl 2-fluorophenyl carboxamide
PubChem CID 4372431
IUPAC Name N-(2,6-dichlorophenyl)-2-fluorobenzamide
SMILES C1=CC=C(C(=C1)C(=O)NC2=C(C=CC=C2Cl)Cl)F

Specifications

Quantity 250 mg
Formula Weight 284.12
Percent Purity 97%
Chemical Name or Material N-(2,6-Dichlorophenyl)-2-fluorobenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.