missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclohexyl-N-methyl-4-nitrobenzamide, 97%, Thermo Scientific™

Product Code. 15426878
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15426878 1 g 1g
15436878 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15426878

Brand: Thermo Scientific Alfa Aesar H60410.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29477-27-8
Molecular Formula C14H18N2O3
Molecular Weight (g/mol) 262.309
MDL Number MFCD00709770
InChI Key ZGDQZPUGCAXWLR-UHFFFAOYSA-N
Synonym benzamide, n-cyclohexyl-n-methyl-4-nitro, aronis24032, n-cyclohexyl-n-methyl 4-nitrophenyl carboxamide
PubChem CID 692212
IUPAC Name N-cyclohexyl-N-methyl-4-nitrobenzamide
SMILES CN(C1CCCCC1)C(=O)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 262.3
Percent Purity 97%
Chemical Name or Material N-Cyclohexyl-N-methyl-4-nitrobenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.