Learn More
NCAM-1/CD56 Antibody (1006411), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals FAB24084F
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
NCAM-1/CD56 Monoclonal antibody specifically detects NCAM-1/CD56 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF
Especificaciones
NCAM-1/CD56 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Simple Western, Flow Cytometry, CyTOF | |
1006411 | |
Western Blot, Simple Western, Flow Cytometry, CyTOF-ready | |
CD56, CD56/ NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM | |
Mouse myeloma cell line NS0-derived recombinant human NCAM-1/CD56, Leu20-Pro603 and Glu636-Asn741, Accession # NP_001070150 | |
0.1 mL | |
Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Membrane Vesicle Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
4684 | |
Store at 4°C in the dark. | |
IgG1 |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Viewing 1-4 of
Título del producto
NCAM-1/CD56 Antibody (1006411), FITC, Novus Biologicals™ > 0.1 mL; FITC
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido