Learn More
Novus Biologicals™ Citrate Buffer Solutions
For recovery of antigens masked by fixation in cross-linking fixatives, such as formalin
Marca: Novus Biologicals™ NB900-66733
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- 10X Citrate buffer for Immunohistochemistry
- Dilute one part buffer with nine parts distilled water
- Unit size: 500mL
Recommended Storage
Store at room temperature
Especificaciones
Temperatura ambiente | |
10x Antigen Retrieval Buffer High pH 10.0 |