Learn More
Löser™ Osmómetros
Operates with low power consumption and no extra water supply is necessary. Loeser™ Freezing Point Osmometer are a cost effective solution for users with smaller numbers of samples.
Marca: Löser™ TYP6
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Freezing initiation by hand
- Cost effective solution for users with smaller numbers of samples.
- Measurements shown on a wide, illuminated LCD display.
- Automatic result recognition and storing ensure reliable measurements
- User guiding and error messages with lamps and display.
- User friendly push button calibration prevents calibration error.
- Third calibration point at 900 mosm provides increased accuracy at higher concentrated solutions.
- Single use plastic tubes.
- Reduced operating costs as no extra water supply is necessary and power consumption is low.
Especificaciones
LCD | |
230 V | |
216 mm | |
Osmómetro | |
Alimentación: contenido de agua en la leche. Hospital: inyecciones de control. Centro de transfusión: control de derivados químicos de la sangre con propiedades fisicoquímicas, y analizados en busca de glicerol residual después de la descongelación. Cultivo celular, bioquímica, biología marina, fisiología de las plantas | |
278 mm | |
230 VCA (100/115 VCA bajo demanda), aproximadamente 45 VA, 50 Hz, 20/45 W | |
1 mOsm/kg | |
20/45 W | |
180 mm |
50 Hz | |
Efecto Peltier | |
1 unidad | |
Tipo 6 | |
Se eleva en concentraciones más altas | |
1.5 min | |
De 0 a 2500 mosm/kg | |
De 50 a 100 μl | |
5.5 kg | |
230 VCA (100/115 VCA bajo demanda), aproximadamente 45 VA, 50 Hz, 20/45 W |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction