missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-boronic acid hydrate, ca 10-15% water, Thermo Scientific™

Product Code. 11348186
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11348186 5 g 5g
11338186 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11348186

Brand: Thermo Scientific Alfa Aesar L15179.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1256355-27-7
Molecular Formula C5H6BNO2
Molecular Weight (g/mol) 122.92
MDL Number MFCD01074545
InChI Key QLULGIRFKAWHOJ-UHFFFAOYSA-N
Synonym pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid
PubChem CID 2734379
IUPAC Name pyridin-4-ylboronic acid
SMILES OB(O)C1=CC=NC=C1

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 471944
Formula Weight 122.92 (Anhydrous)
Assay ≈10 to 15% water
Chemical Name or Material Pyridine-4-boronic acid hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.