missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1,2-Propanediol, 98%, Thermo Scientific Chemicals

Product Code. 11415037
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4254-14-2
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00066248
InChI Key DNIAPMSPPWPWGF-GSVOUGTGSA-N
Synonym r---1,2-propanediol, r-propane-1,2-diol, 2r-propane-1,2-diol, r-1,2-propanediol, r-propylene glycol, 2r-1,2-propanediol, r---1,2-dihydroxypropane, unii-602hn5l69h
PubChem CID 259994
ChEBI CHEBI:28972
IUPAC Name (2R)-propane-1,2-diol
SMILES CC(CO)O

Specifications

Density 1.04
Boiling Point 186°C to 188°C
Flash Point 107°C (224°F)
Refractive Index 1.432
Quantity 5 g
Beilstein 1718872
Sensitivity Hygroscopic
Merck Index 14,7855
Optical Rotation −16.5° (Neat)
Formula Weight 76.1
Percent Purity 98%
Chemical Name or Material (R)-(-)-1,2-Propanediol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'