missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-Methyl-2-pentanol, 99%, Thermo Scientific Chemicals

Product Code. 11339535
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16404-54-9
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD03093077
InChI Key WVYWICLMDOOCFB-ZCFIWIBFSA-N
Synonym r---4-methyl-2-pentanol, 2r-4-methylpentan-2-ol, r-4-methyl-2-pentanol, r-4-methylpentan-2-ol, r---4-methylpentan-2-ol, 4-methyl r-2-pentanol, 2r-4-methyl-2-pentanol, r---methylpentan-2-ol, r--4-methyl-2-pentanol, 2-pentanol, 4-methyl-, 2r
PubChem CID 6999987
IUPAC Name (2R)-4-methylpentan-2-ol
SMILES CC(C)CC(C)O

Specifications

Density 0.802
Boiling Point 130°C to 131°C
Flash Point 41°C (105°F)
Odor Slightly Sweetish
Refractive Index 1.411
Quantity 1 g
UN Number UN2053
Beilstein 1718992
Optical Rotation −21° (Neat)
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material (R)-(-)-4-Methyl-2-pentanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'