missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rb1 Mouse, Clone: RB1/1754, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 5925-MSM2-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Rb1 | |
Nuclear | |
RB1/1754 | |
Unconjugated | |
NP_000312 | |
Mouse | |
105kDa | |
Protein A/G Purified | |
RUO | |
5925 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
RB1 | |
Recombinant human Rb1 protein | |
HeLa, A431 or BT-20 cells. Breast or Colon carcinoma. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |