missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBFOX3 Monoclonal Antibody (OTI2E5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Marca: Origene Technologies TA809952

Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Fox-3, also known as NEUN, HRNBP3, RBFOX3 (RNA binding protein, fox-1) or Fox-1 homolog C, is a 350 amino acid protein that contains one RRM (RNA recognition motif) domain. Localized to both the nucleus and cytoplasm, Fox-3 is suggested to regulate alternative splicing events. Fox-3 is encoded by a gene located on human chromosome 17, which is comprised over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintece of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with maligt cell growth and LiFraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determit of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.Especificaciones
RBFOX3 | |
Monoclonal | |
Unconjugated | |
RBFOX3 | |
FOX-3, FOX3, HRNBP3, NEUN | |
Mouse | |
Affinity Chromatography | |
RUO | |
146713 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI2E5 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
A6NFN3 | |
RBFOX3 | |
Human recombit protein fragment corresponding to amino acids 29-97 of human RBFOX3 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Corrección del contenido de un producto
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
RBFOX3 Monoclonal Antibody (OTI2E5), TrueMAB™, OriGene
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido