Learn More
Elma™ Reaction Tube Holders
Ensure that reaction tubes can be securely positioned and arranged in the Elma Basket.
Marca:  Elma™ 1112158
 Ver más versiones de este producto
Código de producto. 17873094
  • 82.00€ / cada uno
Fecha estimada de envío: 28-03-2025
para ver el stock
Explore más ofertas especiales
Descripción
  • Optimum positioning of the items inside the bath is also a prerequisite for achieving the best possible cleaning results
  • Allows items to be individually positioned
  • Lets you clearly structure your cleaning process
  • Prevents scratches on the tank floor
  • Holders are available for various applications
  • Special solutions are available (e.g. for conical flasks)
Especificaciones
Especificaciones
Reaction Tube Holder
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
100/120