missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacteriophage Fibritin Protein
A DNA sequence encoding the Enterobacteria phage T4 (Bacteriophage T4) Fibritin, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP7036-200ug
Weitere Details : Peso : 0.01000kg
Spezifikation
NP_049771.1 | |
Fibritin Protein | |
Research Use Only | |
wac | |
Recombinant Protein | |
E. coli | |
TDIVLNDLPF VDGPPAEGQS RISWIKNGEE ILGADTQYGS EGSMNRPTVS VLRNVEVLDK NIGILKTSLE TANSDIKTIQ GILDVSGDIE ALAQIGINKK DISDLKTLTS EHTEILNGTN NTVDSILADI GPFNAEANSV YRTIRNDLLW IKRELGQYTG QDINGLPVVG NPSSGMKHRI INNTDVITSQ GIRLSELETK FIESDVGSLT IEVGNLREEL GPKPPSFSQN VYSRLNEIDT KQTTVESDIS AIKTSIGYPG NNSIITSVNT NTDNIASINL ELNQSGGIKQ RLTVIETSIG SDDIPSSIKG QIKDNTTSIE SLNGIVGENT SSGLRANVSW LNQIVGTDSS GGQPSPPGSL LNRVSTIETS VSGLNNAVQN LQVEIGNNSA GIKGQVVALN TLVNGTNPNG STVEERGLTN SIKANETNIA SVTQEVNTAK GNISSLQGDV QALQEAGYIP EAPRDGQAYV RKDGEWVFLS TFLSPA | |
Greater than 90% as determined by SDS-PAGE. |
1258630 | |
200 μg | |
Not determined. | |
Full Length | |
Bacteriophage | |
His-SUMO | |
Tris-based buffer, 50% glycerol |