missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine aFGF Protein

A cDNA sequence encoding the aFGF was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10508-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15944088

  • 212.00€ / 5 microgramos
Fecha estimada de envío: 19-09-2024
para ver el stock

Descripción

Descripción

Especificaciones

Especificaciones

281160
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Stimulates growth of bovine capillary endothelial cells by 3-5 fold over 5% calf serum at 10-25ng/ml FGF and 0ug/ml heparin.
Bovine
Untagged
Each 5microg aFGF were lyophilized from 0.5ml solution containing 1mM sodium phosphate, pH 7 after filtration over a low binding membrane.
aFGF Protein
Research Use Only
FGF1
Recombinant Protein
Bovine
MAEGETTTFT ALTEKFNLPL GNYKKPKLLY CSNGGYFLRI LPDGTVDGTK DRSDQHIQLQ LCAESIGEVY IKSTETGQFL AMDTDGLLYG SQTPNEECLF LERLEENHYN TYISKKHAEK HWFVGLKKNG RSKLGPRTHF GQKAILFLPL PVSSD
Greater than 90%.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales