missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Inositol-1-monophosphatase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Inositol-1-monophosphatase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8609-ec-200ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_417028.1 | |
Inositol-1-monophosphatase Protein | |
Research Use Only | |
ssyA | |
Recombinant Protein | |
E. coli | |
MHPMLNIAVR AARKAGNLIA KNYETPDAVE ASQKGSNDFV TNVDKAAEAV IIDTIRKSYP QHTIITEESG ELEGTDQDVQ WVIDPLDGTT NFIKRLPHFA VSIAVRIKGR TEVAVVYDPM RNELFTATRG QGAQLNGYRL RGSTARDLDG TILATGFPFK AKQYATTYIN IVGKLFNECA DFRRTGSAAL DLAYVAAGRV DGFFEIGLRP WDFAAGELLV REAGGIVSDF TGGHNYMLTG NIVAGNPRVV KAMLANMRDE LSDALKR | |
Greater than 90% as determined by SDS-PAGE. |
947285 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |