missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Coiled-coil domain-containing protein 36 Protein

A DNA sequence encoding the Homo sapiens (Human) Coiled-coil domain-containing protein 36, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7819-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16010991

  • 2385.00€ / 1 miligramo
Fecha estimada de envío: 19-09-2024
para ver el stock

Descripción

Descripción

Especificaciones

Especificaciones

NP_001128669.1
Coiled-coil domain-containing protein 36 Protein
Research Use Only
CCDC36
Recombinant Protein
E. coli
MLSIPSGSGN KKSSNWNNNQ NDYSSLSDSQ FLFGSQFCPE NSETLSAPLD FGAHLRHSKQ SQQNYLEGEP SIFTKYQTKP QLFGGDIKDG GLFPPPLSVG KSKGLLEQFE EKKKRAKDKC DSETLYNFVS NVRESILRLQ TSVEKSEDHL SSRSQSILDS LETVAKTLQE TIQAQNDLVF EAVQDKGNME QAILEMKKRF EARQGEFIEM KSNLKHLEVL VAQQSQEFQQ LCEQLGQLNV PSVLAELKRL ISVPPVKDSA SQTSPPLAQS LNLTRQEKYT SEKPVLWQAQ ALPAAWNPGM GSLQPGEFDV WGEGAKNDDL QEEAALPAFG SHERNRHVKD KVVQTNCKNW AVTKTGAKNH GSSVPGHKIP SDRDLVSQGA SQLTSLEINF STSIKNACQK YQAQSMFLCD PREHLVIKQK DGTVEMRGKD KKQQPRKAHR AHRGRLIASK QKQIPIQTCK FNSKYQSPQP AISVPQSPFL GQQEPRAQPL HLQCPRSPRK PVCPILGGTV MPNKTVRAVQ GRLLQLSRCS SQDNWLLSSS SQGDHQMSWF SDLNLGCSET PLCKEAGKNL LYDLGFDSSD DD
Greater than 90% as determined by SDS-PAGE.
339834
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales