missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Coiled-coil domain-containing protein 36 Protein
A DNA sequence encoding the Homo sapiens (Human) Coiled-coil domain-containing protein 36, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7819-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001128669.1 | |
Coiled-coil domain-containing protein 36 Protein | |
Research Use Only | |
CCDC36 | |
Recombinant Protein | |
E. coli | |
MLSIPSGSGN KKSSNWNNNQ NDYSSLSDSQ FLFGSQFCPE NSETLSAPLD FGAHLRHSKQ SQQNYLEGEP SIFTKYQTKP QLFGGDIKDG GLFPPPLSVG KSKGLLEQFE EKKKRAKDKC DSETLYNFVS NVRESILRLQ TSVEKSEDHL SSRSQSILDS LETVAKTLQE TIQAQNDLVF EAVQDKGNME QAILEMKKRF EARQGEFIEM KSNLKHLEVL VAQQSQEFQQ LCEQLGQLNV PSVLAELKRL ISVPPVKDSA SQTSPPLAQS LNLTRQEKYT SEKPVLWQAQ ALPAAWNPGM GSLQPGEFDV WGEGAKNDDL QEEAALPAFG SHERNRHVKD KVVQTNCKNW AVTKTGAKNH GSSVPGHKIP SDRDLVSQGA SQLTSLEINF STSIKNACQK YQAQSMFLCD PREHLVIKQK DGTVEMRGKD KKQQPRKAHR AHRGRLIASK QKQIPIQTCK FNSKYQSPQP AISVPQSPFL GQQEPRAQPL HLQCPRSPRK PVCPILGGTV MPNKTVRAVQ GRLLQLSRCS SQDNWLLSSS SQGDHQMSWF SDLNLGCSET PLCKEAGKNL LYDLGFDSSD DD | |
Greater than 90% as determined by SDS-PAGE. |
339834 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |