missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human D4GDI Protein
A cDNA sequence encoding the D4GDI was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11594-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
D4GDI Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
D4-GDI protein at 0.1 mg/ml in 50mM Tris-HCl, pH 7.5 and 10mM L-Glutathione (reduced). |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 90% as determined by SDS-PAGE. |