missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dehydrogenase/reductase SDR family member 7 Protein
A DNA sequence encoding the Homo sapiens (Human) Dehydrogenase/reductase SDR family member 7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8296-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001309209.1 | |
Dehydrogenase/reductase SDR family member 7 Protein | |
Research Use Only | |
DHRS7 | |
Recombinant Protein | |
E. coli | |
DGDLTLLWAE WQGRRPEWEL TDMVVWVTGA SSGIGEELAY QLSKLGVSLV LSARRVHELE RVKRRCLENG NLKEKDILVL PLDLTDTGSH EAATKAVLQE FGRIDILVNN GGMSQRSLCM DTSLDVYRKL IELNYLGTVS LTKCVLPHMI ERKQGKIVTV NSILGIISVP LSIGYCASKH ALRGFFNGLR TELATYPGII VSNICPGPVQ SNIVENSLAG EVTKTIGNNG DQSHKMTTSR CVRLMLISMA NDLKEVWISE QPFLLVTYLW QYMPTWAWWI TNKMGKKRIE NFKSGVDADS SYFKIFKTKH D | |
Greater than 90% as determined by SDS-PAGE. |
51635 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |