missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSPBP1 Protein

A cDNA sequence encoding the HSPBP1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12337-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15900109

  • 137.00€ / 2 microgramos
Fecha estimada de envío: 19-09-2024
para ver el stock

Descripción

Descripción

Especificaciones

Especificaciones

HSPBP1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSDEGSRGSR LPLALPPASQ GCSSGGGGGG GGGSSAGGSG NSRPPRNLQG LLQMAITAGS EEPDPPPEPM SEERRQWLQE AMSAAFRGQR EEVEQMKSCL RVLSQPMPPT AGEAEQAADQ QEREGALELL ADLCENMDNA ADFCQLSGMH LLVGRYLEAG AAGLRWRAAQ LIGTCSQNVA AIQEQVLGLG ALRKLLRLLD RDACDTVRVK ALFAISCLVR EQEAGLLQFL RLDGFSVLMR AMQQQVQKLK VKSAFLLQNL LVGHPEHKGT LCSMGMVQQL VALVRTEHSP FHEHVLGALC SLVTDFPQGV RECREPELGL EELLRHRCQL LQQHEEYQEE LEFCEKLLQT CFSSPADDSM DR
Greater than 95% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HSPBP1 solution containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 30% glycerol, 2mM EDTA and 0.1M NaCl.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales