missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL2RG Protein
A cDNA sequence encoding the IL2RG was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12419-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
3561 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
LNTTILTPNG NEDTTADFFL TTMPTDSLSV STLPLPEVQC FVFNVEYMNC TWNSSSEPQP TNLTLHYWYK NSDNDKVQKC SHYLFSEEIT SGCQLQKKEI HLYQTFVVQL QDPREPRRQA TQMLKLQNLV IPWAPENLTL HKLSESQLEL NWNNRFLNHC LEHLVQYRTD WDHSWTEQSV DYRHKFSLPS VDGQKRYTFR VRSRFNPLCG SAQHWSEWSH PIHWGSNTSK ENPFLFALEA HHHHHH | |
Greater than 80.0% by SDS-PAGE. |
IL2RG Protein | |
Research Use Only | |
IL2RG | |
Human | |
His | |
IL2RG protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4). |