Learn More
enQuireBio™ Recombinant Human Leucine-rich repeat neuronal protein 1
A DNA sequence encoding the Homo sapiens (Human) Leucine-rich repeat neuronal protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7749-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001311117.1 | |
Leucine-rich repeat neuronal protein 1 | |
Research Use Only | |
LRRN1 | |
Recombinant Protein | |
E. coli | |
SIQNSECPQL CVCEIRPWFT PQSTYREATT VDCNDLRLTR IPSNLSSDTQ VLLLQSNNIA KTVDELQQLF NLTELDFSQN NFTNIKEVGL ANLTQLTTLH LEENQITEMT DYCLQDLSNL QELYINHNQI STISAHAFAG LKNLLRLHLN SNKLKVIDSR WFDSTPNLEI LMIGENPVIG ILDMNFKPLA NLRSLVLAGM YLTDIPGNAL VGLDSLESLS FYDNKLVKVP QLALQKVPNL KFLDLNKNPI HKIQEGDFKN MLRLKELGIN NMGELVSVDR YALDNLPELT KLEATNNPKL SYIHRLAFRS VPALESLMLN NNALNAIYQK TVESLPNLRE ISIHSNPLRC DCVIHWINSN KTNIRFMEPL SMFCAMPPEY KGHQVKEVLI QDSSEQCLPM ISHDSFPNRL NVDIGTTVFL DCRAMAEPEP EIYWVTPIGN KITVETLSDK YKLSSEGTLE ISNIQIEDSG RYTCVAQNVQ GADTRVATIK VNGTLLDGTQ VLKIYVKQTE SHSILVSWKV NSNVMTSNLK WSSATMKIDN PHITYTARVP VDVHEYNLTH LQPSTDYEVC LTVSNIHQQT QKSCVNVTTK NAAFAVDISD QETSTA | |
Greater than 90% as determined by SDS-PAGE. |
57633 | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |