missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LHRH Protein

A cDNA sequence encoding the LHRH was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12569-HIS-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15900849

  • 212.00€ / 20 microgramos
Fecha estimada de envío: 19-09-2024
para ver el stock

Descripción

Descripción

Especificaciones

Especificaciones

2796
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
Pyr-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2
Greater than 98.0% as determined by RP-HPLC.
LHRH Protein
Research Use Only
GNRH1
Human
His
The LHRH was lyophilized from a concentrated (1 mg/ml) solution with no additives.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales