missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LYG2 Protein

A cDNA sequence encoding the LYG2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12604-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15910969

  • 7875.00€ / 1 miligramo
Fecha estimada de envío: 19-09-2024
para ver el stock

Descripción

Descripción

Especificaciones

Especificaciones

LYG2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSSYPFSHS MKPHLHPRLY HGCYGDIMTM KTSGATCDAN SVMNCGIRGS EMFAEMDLRA IKPYQTLIKE VGQRHCVDPA VIAAIISRES HGGSVLQDGW DHRGLKFGLM QLDKQTYHPV GAWDSKEHLS QATGILTERI KAIQKKFPTW SVAQHLKGGL SAFKSGIEAI ATPSDIDNDF VNDIIARAKF YKRQSF
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The LYG2 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 1mM DTT and 20% glycerol.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales