missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NIT2 Protein

A cDNA sequence encoding the NIT2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12847-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15901739

  • 7875.00€ / 1 miligramo
Fecha estimada de envío: 19-09-2024
para ver el stock

Descripción

Descripción

Especificaciones

Especificaciones

NIT2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMTSFRLA LIQLQISSIK SDNVTRACSF IREAATQGAK IVSLPECFNS PYGAKYFPEY AEKIPGESTQ KLSEVAKECS IYLIGGSIPE EDAGKLYNTC AVFGPDGTLL AKYRKIHLFD IDVPGKITFQ ESKTLSPGDS FSTFDTPYCR VGLGICYDMR FAELAQIYAQ RGCQLLVYPG AFNLTTGPAH WELLQRSRAV DNQVYVATAS PARDDKASYV AWGHSTVVNP WGEVLAKAGT EEAIVYSDID LKKLAEIRQQ IPVFRQKRSD LYAVEMKKP
Greater than 95% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The NIT2 protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 1mM DTT, 0.1M NaCl, and 10% glycerol.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales