missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human PDGF-DD Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marca: R&D Systems™ 1159-SB-025
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Recombinant Human PDGF-DD Protein is derived from NS0. The Recombinant Human PDGF-DD Protein has been validated for the following applications: Bioactivity.Especificaciones
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | |
14 kDa (monomer) | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
Reconstitute at 100μg/mL in sterile 4 mM HCI containing at least 0.1% human or bovine serum albumin. |
80310 | |
PDGF-DD | |
Mouse myeloma cell line,NS0-derived human PDGF-DD protein Ser250-Arg370 | |
Disulfide-linked homodimer | |
Recombinant | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido