missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPM1A / PP2CA / PP2C-alpha Protein
A DNA sequence encoding the Homo sapiens (Human) PPM1A / PP2CA / PP2C-alpha, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6519-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_066283.1 | |
PPM1A / PP2CA / PP2C-alpha Protein | |
Research Use Only | |
PPM1A | |
Recombinant Protein | |
E. coli | |
GAFLDKPKME KHNAQGQGNG LRYGLSSMQG WRVEMEDAHT AVIGLPSGLE SWSFFAVYDG HAGSQVAKYC CEHLLDHITN NQDFKGSAGA PSVENVKNGI RTGFLEIDEH MRVMSEKKHG ADRSGSTAVG VLISPQHTYF INCGDSRGLL CRNRKVHFFT QDHKPSNPLE KERIQNAGGS VMIQRVNGSL AVSRALGDFD YKCVHGKGPT EQLVSPEPEV HDIERSEEDD QFIILACDGI WDVMGNEELC DFVRSRLEVT DDLEKVCNEV VDTCLYKGSR DNMSVILICF PNAPKVSPEA VKKEAELDKY LECRVEEIIK KQGEGVPDLV HVMRTLASEN IPSLPPGGEL ASKRNVIEAV YNRLNPYKND DTDSTSTDDM W | |
Greater than 80% as determined by SDS-PAGE. |
5494 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |