missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Semaphorin-7A Protein
A DNA sequence encoding the Homo sapiens (Human) Semaphorin-7A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9385-ye-10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_001139501.1 | |
Semaphorin-7A Protein | |
Research Use Only | |
SEMA7A | |
Recombinant Protein | |
Yeast | |
QGHLRSGPRI FAVWKGHVGQ DRVDFGQTEP HTVLFHEPGS SSVWVGGRGK VYLFDFPEGK NASVRTVNIG STKGSCLDKR DCENYITLLE RRSEGLLACG TNARHPSCWN LVNGTVVPLG EMRGYAPFSP DENSLVLFEG DEVYSTIRKQ EYNGKIPRFR RIRGESELYT SDTVMQNPQF IKATIVHQDQ AYDDKIYYFF REDNPDKNPE APLNVSRVAQ LCRGDQGGES SLSVSKWNTF LKAMLVCSDA ATNKNFNRLQ DVFLLPDPSG QWRDTRVYGV FSNPWNYSAV CVYSLGDIDK VFRTSSLKGY HSSLPNPRPG KCLPDQQPIP TETFQVADRH PEVAQRVEPM GPLKTPLFHS KYHYQKVAVH RMQASHGETF HVLYLTTDRG TIHKVVEPGE QEHSFAFNIM EIQPFRRAAA IQTMSLDAER RKLYVSSQWE VSQVPLDLCE VYGGGCHGCL MSRDPYCGWD QGRCISIYSS ERSVLQSINP AEPHKECPNP KPDKAPLQKV SLAPNSRYYL SCPMESRHAT YSWRHKENVE QSCEPGHQSP NCILFIENLT AQQYGHYFCE AQEGSYFREA QHWQLLPEDG IMAEHLLGHA CALA | |
Greater than 90% as determined by SDS-PAGE. |
8482 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |