missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human T-complex protein 1 subunit zeta Protein
A DNA sequence encoding the Homo sapiens (Human) T-complex protein 1 subunit zeta, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5780-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001009186.1 | |
T-complex protein 1 subunit zeta Protein | |
Research Use Only | |
CCT6A | |
Recombinant Protein | |
E. coli | |
AAVKTLNPKA EVARAQAALA VNISAARGLQ DVLRTNLGPK GTMKMLVSGA GDIKLTKDGN VLLHEMQIQH PTASLIAKVA TAQDDITGDG TTSNVLIIGE LLKQADLYIS EGLHPRIITE GFEAAKEKAL QFLEEVKVSR EMDRETLIDV ARTSLRTKVH AELADVLTEA VVDSILAIKK QDEPIDLFMI EIMEMKHKSE TDTSLIRGLV LDHGARHPDM KKRVEDAYIL TCNVSLEYEK TEVNSGFFYK SAEEREKLVK AERKFIEDRV KKIIELKRKV CGDSDKGFVV INQKGIDPFS LDALSKEGIV ALRRAKRRNM ERLTLACGGV ALNSFDDLSP DCLGHAGLVY EYTLGEEKFT FIEKCNNPRS VTLLIKGPNK HTLTQIKDAV RDGLRAVKNA IDDGCVVPGA GAVEVAMAEA LIKHKPSVKG RAQLGVQAFA DALLIIPKVL AQNSGFDLQE TLVKIQAEHS ESGQLVGVDL NTGEPMVAAE VGVWDNYCVK KQLLHSCTVI ATNILLVDEI MRAGMSSLKG | |
Greater than 90% as determined by SDS-PAGE. |
908 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |