missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tubulin beta-3 chain Protein
A DNA sequence encoding the Homo sapiens (Human) Tubulin beta-3 chain, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8460-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001184110.1 | |
Tubulin beta-3 chain Protein | |
Research Use Only | |
TUBB3 | |
Recombinant Protein | |
E. coli | |
MREIVHIQAG QCGNQIGAKF WEVISDEHGI DPSGNYVGDS DLQLERISVY YNEASSHKYV PRAILVDLEP GTMDSVRSGA FGHLFRPDNF IFGQSGAGNN WAKGHYTEGA ELVDSVLDVV RKECENCDCL QGFQLTHSLG GGTGSGMGTL LISKVREEYP DRIMNTFSVV PSPKVSDTVV EPYNATLSIH QLVENTDETY CIDNEALYDI | |
Greater than 90% as determined by SDS-PAGE. |
10381 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |