missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H1N1 (Puerto Rico / 8 / 1934) NucleoProtein
A DNA sequence encoding the Influenza A virus (strain A/Puerto Rico/8/1934 H1N1) Influenza A H1N1 (strain A/Puerto Rico/8/1934) Nucleoprotein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7280-ec-1mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_040982.1 | |
Influenza A H1N1 (Puerto Rico / 8 / 1934) NucleoProtein | |
Research Use Only | |
NP | |
Recombinant Protein | |
E. coli | |
MASQGTKRSY EQMETDGERQ NATEIRASVG KMIGGIGRFY IQMCTELKLS DYEGRLIQNS LTIERMVLSA FDERRNKYLE EHPSAGKDPK KTGGPIYRRV NGKWMRELIL YDKEEIRRIW RQANNGDDAT AGLTHMMIWH SNLNDATYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTMV MELVRMIKRG INDRNFWRGE NGRKTRIAYE RMCNILKGKF QTAAQKAMMD QVRESRNPGN AEFEDLTFLA RSALILRGSV AHKSCLPACV YGPAVASGYD FEREGYSLVG IDPFRLLQNS QVYSLIRPNE NPAHKSQLVW MACHSAAFED LRVLSFIKGT KVLPRGKLST RGVQIASNEN METMESSTLE LRSRYWAIRT RSGGNTNQQR ASAGQISIQP TFSVQRNLPF DRTTIMAAFN GNTEGRTSDM RTEIIRMMES ARPEDVSFQG RGVFELSDEK AASPIVPSFD MSNEGSYFFG DNAEEYDN | |
Greater than 80% as determined by SDS-PAGE. |
956531 | |
1 mg | |
Not determined. | |
Full Length | |
Influenza | |
His-B2M | |
Tris-based buffer, 50% glycerol |