missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL13 / ALRH Protein
A DNA sequence encoding the Rhesus Macaque IL13 / ALRH, was expressed in the hosts and tags indicated.
105.00€ - 4105.00€
Especificaciones
Número de acceso | NP_001028101.1 |
---|---|
ID de gen (Entrez) | 574325 |
Nombre | IL13 / ALRH Protein |
Estado normativo | Research Use Only |
Concentración de endotoxinas | Less than 1.0 EU/ug as determined by LAL method. |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15943868
|
enQuireBio™
QP10442-ec-2ug |
2 μg | |||||||
15923868
|
enQuireBio™
QP10442-ec-10ug |
10 μg | |||||||
15913868
|
enQuireBio™
QP10442-ec-100ug |
100 μg | |||||||
15933868
|
enQuireBio™
QP10442-ec-250ug |
250 μg | |||||||
Especificaciones
NP_001028101.1 | |
IL13 / ALRH Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SPSPVPRSTA LKELIEELVN ITQNQKAPLC NGSMVWSINL TAGVYCAALE SLINVSGCSA IEKTQRMLNG FCPHKVSAGQ FSSLRVRDTK IEVAQFVKDL LVHLKKLFRE GRFN | |
Greater than 98% as determined by SDS-PAGE and HPLC. |
574325 | |
Research Use Only | |
IL13 | |
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 5 ng/ml, corresponding to a specific activity of > 2.0 M 105 IU/mg. | |
Macaque | |
Untagged | |
0.2 um filtered PBS, pH 7.4, 3% trehalose, lyophilized |