Learn More
R&D Systems™ Recombinant Mouse FGF-6 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marca:  R&D Systems™ 5750-F6-025
 Ver más versiones de este producto
Código de producto. 18186480
  • 348.00€ / 25 microgramos
Explore más ofertas especiales
Descripción
Descripción
The Recombinant Mouse FGF-6 Protein is derived from E. coli. The Recombinant Mouse FGF-6 Protein has been validated for the following applications: Bioactivity.
Especificaciones
Especificaciones
Lyophilized from a 0.2μm filtered solution in MOPS, Ammonium Sulfate, EDTA, DTT with BSA as a carrier protein.
18.8 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
14177
FGF-6
E. coli-derived mouse FGF-6 protein Pro39-Ile208
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.