Learn More
enQuireBio™ Recombinant other Deslorelin Protein
Recombinant Protein
Marca:  enQuireBio™ QP11661-25mg
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15997948
  • 301.00€ / 25 miligramos
Fecha estimada de envío: 19-09-2024
para ver el stock
Especificaciones
Deslorelin Protein
Research Use Only
Recombinant Protein
E. coli
The Deslorelin peptide was lyophilized with no additives.
25 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Greater than 99.0% as determined by RP-HPLC.