missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit IGF1 Protein
A cDNA sequence encoding the IGF1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10679-50ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
IGF1 Protein | |
Research Use Only | |
IGF-I is biologically active when compared to human IGF-1. The ED50, calculated by the dose -dependent proliferation of human MCF/7 cells is 5 to 25ng/ml in the cell culture mixture dependent on culture conditions. Its activity consists of 30-40 % compared to human IGF-1. | |
Rabbit | |
Untagged | |
The protein was lyophilized from a concentrated (0.5 mg/ml) solution with 0.02% NaHCO3. |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MTAGPETLCG AELVDALQFV CGDRGFYFNK PTGYGSSSRR APQTGIVDEC CFRSCDLRRL EMYCAPLKP | |
Greater than 98.0% as determined by:(a) Gel-filtration chromatography under non denaturing conditions. (b) Analysis by SDS-PAGE. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido