missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat NME2 Protein

A cDNA sequence encoding the NME2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12854-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15901759

  • 4090.00€ / 1 miligramo
Fecha estimada de envío: 19-09-2024
para ver el stock

Descripción

Descripción

Especificaciones

Especificaciones

NME2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMANLERT FIAIKPDGVQ RGLVGEIIKR FEQKGFRLVA MKFLRASEEH LKQHYIDLKD RPFFPGLVKY MNSGPVVAMV WEGLNVVKTG RVMLGETNPA DSKPGTIRGD FCIQVGRNII HGSDSVESAE KEIGLWFKPE ELIDYKSCAH DWVYE
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Rat
His
NME2 protein solution (1 mg/ml) containing Phosphate Buffered Saline pH 7.4 and 10% glycerol.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales