missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Toxoplasma Gondii T. gondii p29 Protein
A cDNA sequence encoding the T. gondii p29 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13665-500ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Toxoplasma Gondii T. gondii p29 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, 1.5M urea and 50% glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Toxoplasma Gondii | |
GST | |
Toxoplasma protein is >95% pure as determined by 10% PAGE (coomassie staining). |