missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Rhesus macaque EGF R Protein, His Tag

Marca:  ACROBiosystems EGR-C52H1-50UG

Detalles adicionales : Peso : 0.00001kg

 Ver más versiones de este producto

Código de producto. 15886617

  • 302.42€ / 50 microgramos
Fecha estimada de envío: 07-08-2024
para ver el stock

Descripción

Descripción

The epidermal growth factor receptor (EGFR; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. The epidermal growth factor receptor is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: EGFR (ErbB-1), HER2/c-neu (ErbB-2), Her 3 (ErbB-3) and Her 4 (ErbB-4). Mutations affecting EGFR expression or activity could result in cancer.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.5 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones

Especificaciones

70.5 kDa
Rhesus macaque EGF R, His Tag (EGR-C52H1) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P55245).
-20°C
EGF R
Rhesus Macaque
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens
50 μg
Leu 25 - Ser 645
EGFR,ERBB,ERBB1,HER1,PIG61,mENA
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales