Learn More
Elma™ Rinsing Sinks
This rinsing sink removes any remaining contamination after cleaning.
Marca:  Elma™ 1113841
 Ver más versiones de este producto
Código de producto. 17821307
  • 287.00€ / cada uno
Explore más ofertas especiales
Descripción
Descripción
  • Important addition to the overall cleaning process
  • Tabletop unit is available in four different sizes – the L and XL models feature an additional side drain
  • Once the cleaning process in the ultrasonic unit is complete, the basket can be inserted directly into the Elma Sink
  • Ensures that any residues and cleaning chemicals are rinsed off thoroughly
  • In conjunction with a suitable rinsing medium, this guarantees optimal cleaning results
Especificaciones
Especificaciones
Sink
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
L