Learn More
Elma™ Rinsing Sinks
This rinsing sink removes any remaining contamination after cleaning.
Marca: Elma™ 1113841
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Important addition to the overall cleaning process
- Tabletop unit is available in four different sizes – the L and XL models feature an additional side drain
- Once the cleaning process in the ultrasonic unit is complete, the basket can be inserted directly into the Elma Sink
- Ensures that any residues and cleaning chemicals are rinsed off thoroughly
- In conjunction with a suitable rinsing medium, this guarantees optimal cleaning results
Especificaciones
Sink | |
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY | |
L |