Learn More
Hettich Lab Technology™ Slide Carrier
Product Code. 12003894
Click to view available options
Capacity:
1, 2, 4, 8 mL, 3 x 2 mL, 4 x 1 mL
2 x 1, 2, 4, 8 mL, 3 x 2 mL, 4 x 1 mL
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 12003894
Brand: Hettich Lab Technology™ 1670
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Hettich Lab Technology™ Slide Carrier is available as a single or double chamber slide/filter carrier for cyto-centrifuge systems.
  • For cyto centrifuge systems and cell evaluations
  • Accepts funneling cyto chambers 1663-1668
Specifications
Type Slide Carrier
For Use With (Equipment) Cyto centrifuge system, Filter cards 1691-1694, Funneling cyto chambers 1663-1668
Capacity 2 x 1, 2, 4, 8 mL, 3 x 2 mL, 4 x 1 mL
Autoclavable Autoclavable
Includes 2 Adapter Rings
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.