missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetracyanoplatinate(II) hydrate, Premion™, 99.95% (metals basis), Pt 48.4% min, Thermo Scientific Chemicals

Product Code. 11303998
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11303998

Brand: Thermo Scientific Alfa Aesar 040400.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 699012-94-7
Molecular Formula C4N4Na2Pt
Molecular Weight (g/mol) 345.13
MDL Number MFCD00798527
InChI Key WOIFBVMIVVRIIE-UHFFFAOYSA-N
Synonym disodium tetracyanoplatinumdiuide hydrate, sodium tetracyanoplatinate ii hydrate, premion
PubChem CID 16217382
IUPAC Name disodium tetracyanoplatinumbis(ylium)
SMILES [Na+].[Na+].N#C[Pt++](C#N)(C#N)C#N

Specifications

Melting Point (decomposition)
Color White
Physical Form Crystalline Powder
Quantity 5 g
Formula Weight 345.14 (Anhydrous)
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Pt 48.4% min.
Assay (metals basis)
Chemical Name or Material Sodium tetracyanoplatinate(II) hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.