Learn More
Thermo Scientific™ SUN-SRi™ SunFlo™ Dissolution Filters
Manufactured of Ultra High Molecular Weight (UHMW) foamed porous polyethylene
Marca: Thermo Scientific™ 14823272
Detalles adicionales : Peso : 0.80000kg
Descripción
- Full flow design removes small particulates
- Fit securely on the end of a 1/8 in. cannula or sample probe
Especificaciones
45 μm | |
Case of 1000 | |
1/8 in. Cannula or Probe |