missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL Sylicon Conical Stopper
The Sylicon Conical Stopper creates a seal in bottles and flasks.
Marca: KARTELL 0381200
Detalles adicionales : Peso : 0.10000kg
Descripción
- Autoclavable.
- SUITABLE FOR FOODSTUFF.
- MATERIAL: SYLICON
Especificaciones
Non-sterile | |
32 mm | |
Silicone | |
42 mm |
White | |
42 mm | |
Autoclavable |