Learn More
Syntaxin 4 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-32227JF646
Descripción
Syntaxin 4 Polyclonal antibody specifically detects Syntaxin 4 in Human samples. It is validated for Western Blot
Especificaciones
Syntaxin 4 | |
Polyclonal | |
Western Blot | |
Renal carcinoma antigen NY-REN-31, STX4Ap35-2, syntaxin 4, syntaxin 4A (placental), syntaxin-4 | |
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Syntaxin 4 using the numbering given in entry NP_004595.2 (GeneID 6810). | |
0.1 mL | |
Neuroscience, Signal Transduction | |
6810 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.