Learn More
Invitrogen™ Vitronectin (VTN) Human ELISA Kit
Sandwich ELISA Kit
634.00€ - 2635.00€
Especificaciones
Número de acceso | P04004 |
---|---|
Sensibilidad del ensayo | 0,8 ng |
ensayo | ELISA sandwich de fase sólida |
Conjugado | Biotina |
Tipo de producto | Kit de ELISA |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15465173
|
Invitrogen™
EHVTN |
96 pruebas |
634.00€
1 set |
Fecha estimada de envío: 21-08-2024
Inicie sesión para ver el stock |
|||||
17175599
|
Invitrogen™
EHVTNX5 |
5 × 96 pruebas |
2635.00€
cada uno |
Fecha estimada de envío: 28-08-2024
Inicie sesión para ver el stock |
|||||
Descripción
The Human Vitronectin (VTN) ELISA quantitates Hu VTN in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu VTN. Principle of the method The Human VTN solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Vitronectin is a plasma glycoprotein that circulates in the blood. Vitronectin is circulating as a mixture of both 75 kDa and 65 kDa forms. Vitronectin is a major cell adhesive glycoprotein and is a common component of extracellular matrix and plasma. It competes effectively with other plasma proteins and is often involved in cell attachment, regulation of blood coagulation and immune responses. It has similar tissue distribution to fibronectin and also its integrin receptor recognises fibronectin.Especificaciones
P04004 | |
ELISA sandwich de fase sólida | |
Kit de ELISA | |
Plasma, Sobrenadante, Suero | |
ELISA, ensayos de proteínas, biología de proteínas, biología del cáncer, inmunología, neurociencia, neurología, detección de proteínas, análisis de proteínas | |
7448 | |
< 12 % | |
Peroxidasa de rábano picante (HRP) | |
Investigación cardiovascular | |
V75, VN, VNT | |
1 h 20 min |
0,8 ng | |
Biotina | |
Otras proteínas | |
Humano | |
Lector de microplacas colorimétrico | |
V75,VN,VNT | |
< 10 % | |
RUO | |
De 2 °C a 8 °C | |
Humano | |
4 h 45 min |