CAS RN 1115-70-4
CAS RN 1115-70-4
IUPAC Name:
hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride
Sinónimos:
hydrogen metformin (inn) and its salts chloride
Peso molecular (g/mol):
165.63
Fórmula molecular:
C4H12ClN5
InChi Key:
OETHQSJEHLVLGH-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CN(C)C(=N)N=C(N)N
1
–
5
de
5
resultados
2
MetFormin hydrochloride, 98%, For HPLC analysis, MP Biomedicals™
CAS: 1115-70-4 Fórmula molecular: C4H12ClN5 Peso molecular (g/mol): 165.63 Número MDL: MFCD00012582 Clave InChI: OETHQSJEHLVLGH-UHFFFAOYSA-N Sinónimo: metformin hydrochloride,1,1-dimethylbiguanide hydrochloride,metformin hcl,glucophage,riomet,diabefagos,meguan,metformin monohydrochloride,benofomin,denkaform PubChem CID: 14219 Nombre IUPAC: Hidrógeno 1-[(diaminometilideno)amino]-N,N-dimetilmetanimidamida cloruro SMILES: [H+].[Cl-].CN(C)C(=N)N=C(N)N
![EDGE](/content/dam/fishersci/glyphs/edge.png)
5
STEMCELL Technologies Metformin (Hydrochloride)
AMPK activator; Mitochondrial respiratory chain complex I inhibitor
![ENCOMPASS_PREFERRED](/content/dam/fishersci/glyphs/encompass_preferred.png)