missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)isoquinoline hydrobromide, 97%, Thermo Scientific™

Product Code. 10461684
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 337508-56-2
Molecular Formula C10H9Br2N
Molecular Weight (g/mol) 302.997
MDL Number MFCD03086096
InChI Key RWSWRJTWPXSENX-UHFFFAOYSA-N
Synonym 1-bromomethyl isoquinoline hydrobromide, 1-bromomethylisoquinoline hydrobromide, isoquinoline, 1-bromomethyl-, hydrobromide 1:1, 1-bromomethylisoquinoline hbr, 1-bromomethyl-isoquinolin hydrobromide, 1-bromomethyl-isoquinoline hydrobromide, isoquinoline, 1-bromomethyl-, hydrobromide
PubChem CID 2776245
IUPAC Name 1-(bromomethyl)isoquinoline;hydrobromide
SMILES C1=CC=C2C(=C1)C=CN=C2CBr.Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 303
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)isoquinoline hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(Bromomethyl)isoquinoline hydrobromide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.